accessibility menu, dialog, popup

UserName

CAS RN 16871-90-2

KKFSiFFFFF

CAS RN 16871-90-2

IUPAC Name: dipotassium hexafluorosilanediuide
Synonyms: dipotassium hexafluorosilanediuide
Molecular Weight (g/mol): 220.27
Molecular Formula: F6K2Si
InChi Key: UUHPPUWEDCVPCO-UHFFFAOYSA-N
SMILES: [K+].[K+].F[Si--](F)(F)(F)(F)F
Physical Form 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)