accessibility menu, dialog, popup

UserName

CAS RN 16830-15-2

H3C(R)(S)(S)CH3(R)H3C(R)OH(S)OH(R)H3COH(R)CH3(S)(S)CH3OOOOOHOOOCH3HOHOHOHOOHHOOHOH

CAS RN 16830-15-2

IUPAC Name: 6-({[3,4-dihydroxy-6-(hydroxymethyl)-5-[(3,4,5-trihydroxy-6-methyloxan-2-yl)oxy]oxan-2-yl]oxy}methyl)-3,4,5-trihydroxyoxan-2-yl (1S,2R,4aS,6aS,6bR,9R,10S,11R,12aR,14bS)-10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylate
Synonyms: asiaticoside
Molecular Weight (g/mol): 959.13
Molecular Formula: C48H78O19
InChi Key: WYQVAPGDARQUBT-NNUNPJGWSA-N
SMILES: C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CCC4[C@@]5(C)C[C@@H](O)[C@@H](O)[C@@](C)(CO)C5CC[C@@]34C)[C@@H]2[C@H]1C)C(=O)OC1OC(COC2OC(CO)C(OC3OC(C)C(O)C(O)C3O)C(O)C2O)C(O)C(O)C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results