CAS RN 1675248-18-6
CAS RN 1675248-18-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Descyano-2-ethyl Formyl-tofacitinib, TRC
CAS: 1675248-18-6 Molecular Formula: C18 H25 N5 O3 Molecular Weight (g/mol): 359.423 Synonym: Ethyl 3-((3R,4R)-4-Methyl-3-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)piperidin-1-yl)-3-oxopropanoate IUPAC Name: ethyl 3-[(3R,4R)-4-methyl-3-[methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]piperidin-1-yl]-3-oxopropanoate SMILES: CCOC(=O)CC(=O)N1CC[C@@H](C)[C@H](C1)N(C)c2ncnc3[nH]ccc23