accessibility menu, dialog, popup

UserName

ONOFBrF

CAS RN 167415-27-2

IUPAC Name: 1-bromo-2,5-difluoro-4-nitrobenzene
Synonyms: 1-bromo-2,5-difluoro-4-nitrobenzene
Molecular Weight (g/mol): 237.99
Molecular Formula: C6H2BrF2NO2
InChi Key: GJFYMYJYPARISZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(Br)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results