CAS RN 167145-13-3
IUPAC Name:
2-[2-(3-methoxyphenyl)ethyl]phenol
Synonyms:
2-[2-(3-methoxyphenyl)ethyl]phenol
Molecular Weight (g/mol):
228.29
Molecular Formula:
C15H16O2
InChi Key:
HGQQRAXOBYWKDV-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(CCC2=CC=CC=C2O)=C1
1
–
1
of
1
results
1
2-[2-(3-Methoxyphenyl)ethyl]phenol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.