accessibility menu, dialog, popup

UserName

OOOOOOOOOOOOH2OH2OH2OH2OH2OH2OFeFe

CAS RN 166897-40-1

IUPAC Name: diiron(3+) hexahydrate trioxalate
Synonyms: diiron(3+) hexahydrate trioxalate
Molecular Weight (g/mol): 483.83
Molecular Formula: C6H12Fe2O18
InChi Key: FWXIZVVTJVNNRX-UHFFFAOYSA-H
SMILES: O.O.O.O.O.O.[Fe+3].[Fe+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results