accessibility menu, dialog, popup

UserName

CH3NNHOHNNOO(E/Z)

CAS RN 165252-70-0

IUPAC Name: N''-methyl-N-nitro-N'-[(oxolan-3-yl)methyl]guanidine
Synonyms: dinotefuran
Molecular Weight (g/mol): 202.21
Molecular Formula: C7H14N4O3
InChi Key: YKBZOVFACRVRJN-UHFFFAOYNA-N
SMILES: CN=C(NCC1CCOC1)N[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results