accessibility menu, dialog, popup

UserName

CH3H3CHOOHOHNOH

CAS RN 16485-10-2

IUPAC Name: 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Synonyms: panthenol 泛醇 panthenol [usan:ban:inn:jan]
Molecular Weight (g/mol): 205.25
Molecular Formula: C9H19NO4
InChi Key: SNPLKNRPJHDVJA-UHFFFAOYNA-N
SMILES: CC(C)(CO)C(O)C(=O)NCCCO
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results