accessibility menu, dialog, popup

UserName

CAS RN 16387-71-6

CH3H3C

CAS RN 16387-71-6

IUPAC Name: deca-4,6-diyne
Synonyms: deca-4,6-diyne
Molecular Weight (g/mol): 134.22
Molecular Formula: C10H14
InChi Key: LIWZSNTUMSGWTF-UHFFFAOYSA-N
SMILES: CCCC#CC#CCCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results