accessibility menu, dialog, popup

UserName

CAS RN 161622-14-6

IUPAC Name: 4-bromo-3-(trifluoromethyl)benzoic acid
Synonyms: 4-bromo-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 269.02
Molecular Formula: C8H4BrF3O2
InChi Key: GPBPFDPENZHCPR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=C(Br)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results