accessibility menu, dialog, popup

UserName

CH3CH3NOON

CAS RN 16034-49-4

IUPAC Name: 2-(3,5-dimethyl-1H-pyrazol-1-yl)acetate
Synonyms: 2-(3,5-dimethylpyrazol-1-yl)acetate
Molecular Weight (g/mol): 153.16
Molecular Formula: C7H9N2O2
InChi Key: JYSWEDYPQJOEPO-UHFFFAOYSA-M
SMILES: CC1=NN(CC([O-])=O)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results