accessibility menu, dialog, popup

UserName

CAS RN 15764-16-6

CH3OH3C

CAS RN 15764-16-6

IUPAC Name: 2,4-dimethylbenzaldehyde
Synonyms: 2,4-dimethylbenzaldehyde
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: GISVICWQYMUPJF-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=O)C(C)=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)