CAS RN 157283-66-4
CAS RN 157283-66-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isopropyl Cloprostenate, TRC
CAS: 157283-66-4 Molecular Formula: C25H35ClO6 Molecular Weight (g/mol): 466.99 Synonym: (5Z)-7-[(1R,2R,3R,5S)-2-[(1E,3R)-4-(3-Chlorophenoxy)-3-hydroxy-1-buten-1-yl]-3,5-dihydroxycyclopentyl]-5-heptenoic Acid 1-Methylethyl Ester,[1R-[1alpha(Z),2beta(1E,3R*),3alpha,5alpha]]-7-[2-[4-(3-Chlorophenoxy)-3-hydroxy-1-butenyl]-3,5-dihydroxycyclopentyl]-5-heptenoic Acid 1-Methylethyl Ester,CPG 6112-95,Cloprostenol Isopropyl Ester SMILES: CC(C)OC(=O)CCC\C=C/C[C@H]1[C@@H](O)C[C@@H](O)[C@@H]1\C=C\[C@@H](O)COc2cccc(Cl)c2