accessibility menu, dialog, popup

UserName

CAS RN 15707-23-0

H3CNNH3C

CAS RN 15707-23-0

IUPAC Name: 2-ethyl-3-methylpyrazine
Synonyms: 2-ethyl-3-methylpyrazine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: LNIMMWYNSBZESE-UHFFFAOYSA-N
SMILES: CCC1=NC=CN=C1C
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)