accessibility menu, dialog, popup

UserName

CAS RN 15572-79-9

HO(S)O(R)OH(S)OH(R)HO(S)HO

CAS RN 15572-79-9

IUPAC Name: (2R,3S,4R,5S,6S)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Synonyms: α-L-galactose
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: WQZGKKKJIJFFOK-SXUWKVJYSA-N
SMILES: OC[C@@H]1O[C@@H](O)[C@@H](O)[C@H](O)[C@@H]1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results