CAS RN 1544-86-1
IUPAC Name:
1-(difluoromethoxy)-4-nitrobenzene
Synonyms:
1-(difluoromethoxy)-4-nitrobenzene
Molecular Weight (g/mol):
189.12
Molecular Formula:
C7H5F2NO3
InChi Key:
SVGGBARCOQPYMV-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=C(OC(F)F)C=C1
1
–
1
of
1
results
1
1-Difluoromethoxy-4-nitrobenzene, 98%, Thermo Scientific™
CAS: 1544-86-1 Molecular Formula: C7H5F2NO3 Molecular Weight (g/mol): 189.118 MDL Number: MFCD00085009 InChI Key: SVGGBARCOQPYMV-UHFFFAOYSA-N Synonym: 4-difluoromethoxy nitrobenzene,1-difluoromethoxy-4-nitrobenzene,1-difluoromethoxy-4-nitro-benzene,4-difluoromethoxy-1-nitrobenzene,acmc-209dar,4-difluoromethoxynitrobenzene,svggbarcoqpymv-uhfffaoysa,4-nitro-alpha,alpha-difluoroanisole PubChem CID: 2737014 IUPAC Name: 1-(difluoromethoxy)-4-nitrobenzene SMILES: C1=CC(=CC=C1[N+](=O)[O-])OC(F)F