CAS RN 154026-95-6
CAS RN 154026-95-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tert-butyl 2-[(4R,6S)-2,2-Dimethyl-6-[(methylcarbonyloxy)methyl]-1,3-dioxan-4-yl]acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Tert-butyl 2-[(4R,6S)-2,2-Dimethyl-6-[(methylcarbonyloxy)methyl]-1,3-dioxan-4-yl]acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tert-butyl 2-[(4R,6S)-2,2-Dimethyl-6-[(methylcarbonyloxy)methyl]-1,3-dioxan-4-yl]acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.