accessibility menu, dialog, popup

UserName

CAS RN 15307-93-4

ClClHN

CAS RN 15307-93-4

IUPAC Name: 2,6-dichloro-N-phenylaniline
Synonyms: 2,6-dichloro-N-phenylaniline
Molecular Weight (g/mol): 238.11
Molecular Formula: C12H9Cl2N
InChi Key: HDUUZPLYVVQTKN-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(Cl)=C1NC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results