accessibility menu, dialog, popup

UserName

CAS RN 15190-10-0

NNO

CAS RN 15190-10-0

IUPAC Name: 2-(morpholin-4-yl)-2-phenylacetonitrile
Synonyms: 2-(morpholin-4-yl)-2-phenylacetonitrile
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: MWZPYQLYZXTCLZ-UHFFFAOYNA-N
SMILES: N#CC(N1CCOCC1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results