CAS RN 151414-47-0
CAS RN 151414-47-0
IUPAC Name:
2,6-difluoro-4-methoxyaniline
Synonyms:
2,6-difluoro-4-methoxyaniline
Molecular Weight (g/mol):
159.14
Molecular Formula:
C7H7F2NO
InChi Key:
HQBURSNJKAITGQ-UHFFFAOYSA-N
SMILES:
COC1=CC(F)=C(N)C(F)=C1
1
–
1
of
1
results
1
2,6-Difluoro-4-methoxyaniline, 97%, Thermo Scientific™
CAS: 151414-47-0 Molecular Formula: C7H7F2NO Molecular Weight (g/mol): 159.136 MDL Number: MFCD04115922 InChI Key: HQBURSNJKAITGQ-UHFFFAOYSA-N Synonym: 4-amino-3,5-difluoroanisole,2,6-difluoro-4-methoxyphenylamine,benzenamine, 2,6-difluoro-4-methoxy,2,6-difluoro-4-methoxy-benzenamine,2,6-difluoro-4-methoxy-phenylamine,intermediates-zcf02157,acmc-209z8h,4-methoxy-2,6-difluoroaniline,2,6-difluoro-4-methoxybenzenamine,benzenamine,2,6-difluoro-4-methoxy PubChem CID: 5185763 IUPAC Name: 2,6-difluoro-4-methoxyaniline SMILES: COC1=CC(=C(C(=C1)F)N)F