CAS RN 1506-53-2
CAS RN 1506-53-2
IUPAC Name:
N-dodecylprop-2-enamide
Synonyms:
N-dodecylprop-2-enamide
Molecular Weight (g/mol):
239.4
Molecular Formula:
C15H29NO
InChi Key:
XQPVIMDDIXCFFS-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCNC(=O)C=C
1
–
1
of
1
results
1
N-Dodecylacrylamide, 98+%, Thermo Scientific™
CAS: 1506-53-2 Molecular Formula: C15H29NO Molecular Weight (g/mol): 239.40 MDL Number: MFCD08276361 InChI Key: XQPVIMDDIXCFFS-UHFFFAOYSA-N Synonym: n-dodecylacrylamide,2-propenamide,n-dodecyl,acmc-209d4r PubChem CID: 73929 IUPAC Name: N-dodecylprop-2-enamide SMILES: CCCCCCCCCCCCNC(=O)C=C