accessibility menu, dialog, popup

UserName

CAS RN 150374-95-1

NaH3CH3CCH3OOSOONHOHNOO

CAS RN 150374-95-1

IUPAC Name: sodium 2-[(2-{4-[(2,2-dimethylpropanoyl)oxy]benzenesulfonamido}phenyl)formamido]acetate
Synonyms: sodium 2-[(2-{4-[(2,2-dimethylpropanoyl)oxy]benzenesulfonamido}phenyl)formamido]acetate
Molecular Weight (g/mol): 456.44
Molecular Formula: C20H21N2NaO7S
InChi Key: ZAIFANJZUGNYCK-UHFFFAOYSA-M
SMILES: [Na+].CC(C)(C)C(=O)OC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=C1C(=O)NCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results