accessibility menu, dialog, popup

UserName

CAS RN 149597-92-2

H3N(S)OO

CAS RN 149597-92-2

IUPAC Name: (2S)-2-azaniumyl-3,3-diphenylpropanoate
Synonyms: diphenylalanine
Molecular Weight (g/mol): 241.29
Molecular Formula: C15H15NO2
InChi Key: PECGVEGMRUZOML-AWEZNQCLSA-N
SMILES: [NH3+][C@@H](C(C1=CC=CC=C1)C1=CC=CC=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)