accessibility menu, dialog, popup

UserName

CAS RN 149-87-1

OHOOHN

CAS RN 149-87-1

IUPAC Name: 5-oxopyrrolidine-2-carboxylic acid
Synonyms: pyroglutamate acidum pidolicum pidolate, magnesium DL-pyroglutamic acid L-glutiminate acid, pyrrolidonecarboxylic glutiminate ajidew A-100 (S)-pyroglutamate glutimate
Molecular Weight (g/mol): 129.12
Molecular Formula: C5H7NO3
InChi Key: ODHCTXKNWHHXJC-UHFFFAOYNA-N
SMILES: OC(=O)C1CCC(=O)N1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)