CAS RN 146000-39-7
CAS RN 146000-39-7
IUPAC Name:
2-{[(tert-butoxy)carbonyl](methyl)amino}-2-methylpropanoate
Synonyms:
2-[(tert-butoxycarbonyl)(methyl)amino]-2-methylpropanoate
Molecular Weight (g/mol):
216.26
Molecular Formula:
C10H18NO4
InChi Key:
KPDLITGXUYMJEC-UHFFFAOYSA-M
SMILES:
CN(C(=O)OC(C)(C)C)C(C)(C)C([O-])=O
1
–
1
of
1
results
1
N-BOC-N,2-Dimethylalanine, 98+%
CAS: 146000-39-7 Molecular Formula: C10H19NO4 Molecular Weight (g/mol): 217.27 MDL Number: MFCD01862912 InChI Key: KPDLITGXUYMJEC-UHFFFAOYSA-N Synonym: boc-n-me-aib-oh,boc-n,2-dimethylalanine,boc-n-methyl-aminoisobutyric acid,2-tert-butoxycarbonyl methyl amino-2-methylpropanoic acid,boc-n-methyl-aib,acmc-1c0qg,n-boc-n,2-dimethylalanine,n-boc-n,2-dimethyl-dl-alanine,2-methyl-2-methyl-2-methylpropan-2-yl oxycarbonyl amino propanoic acid,n-boc-,a-methylamino isobutyric acid PubChem CID: 2734752 IUPAC Name: 2-methyl-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid SMILES: CN(C(=O)OC(C)(C)C)C(C)(C)C(O)=O