accessibility menu, dialog, popup

UserName

CAS RN 145084-28-2

CH3NHONH(R)NNOCH3O

CAS RN 145084-28-2

IUPAC Name: 3-(3-methylphenyl)-1-[(3R)-1-[2-(2-methylphenyl)-2-oxoethyl]-2-oxo-5-phenyl-2,3-dihydro-1H-1,4-benzodiazepin-3-yl]urea
Synonyms: 3-(3-methylphenyl)-1-[(3R)-1-[2-(2-methylphenyl)-2-oxoethyl]-2-oxo-5-phenyl-3H-1,4-benzodiazepin-3-yl]urea
Molecular Weight (g/mol): 516.6
Molecular Formula: C32H28N4O3
InChi Key: YCXFHPUBGMMWJQ-PMERELPUSA-N
SMILES: CC1=CC=CC(NC(=O)N[C@@H]2N=C(C3=CC=CC=C3)C3=CC=CC=C3N(CC(=O)C3=CC=CC=C3C)C2=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results