accessibility menu, dialog, popup

UserName

CAS RN 1449132-59-5

BOHNFFClHOOH

CAS RN 1449132-59-5

IUPAC Name: {2-chloro-5-[(2,5-difluorophenyl)carbamoyl]phenyl}boronic acid
Synonyms: 2-chloro-5-[(2,5-difluorophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol): 311.48
Molecular Formula: C13H9BClF2NO3
InChi Key: OPNXXIVOUWDWMG-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(=C1)C(=O)NC1=C(F)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results