accessibility menu, dialog, popup

UserName

BONH3CFHOOH

CAS RN 1449132-53-9

IUPAC Name: [3-fluoro-4-(4-methylpiperidine-1-carbonyl)phenyl]boronic acid
Synonyms: 3-fluoro-4-(4-methylpiperidine-1-carbonyl)phenylboronic acid
Molecular Weight (g/mol): 265.09
Molecular Formula: C13H17BFNO3
InChi Key: NTDBICSBLSVBSL-UHFFFAOYSA-N
SMILES: CC1CCN(CC1)C(=O)C1=C(F)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results