accessibility menu, dialog, popup

UserName

CAS RN 1449132-27-7

OOHHNSOOCl

CAS RN 1449132-27-7

IUPAC Name: 2-(3-chlorobenzenesulfonamido)-3-phenylpropanoic acid
Synonyms: 2-(3-chlorobenzenesulfonamido)-3-phenylpropanoic acid
Molecular Weight (g/mol): 339.79
Molecular Formula: C15H14ClNO4S
InChi Key: XZBAIPMBXLSRKT-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=CC=C1)NS(=O)(=O)C1=CC(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)