accessibility menu, dialog, popup

UserName

CAS RN 14404-33-2

ClRuClClClClKK

CAS RN 14404-33-2

IUPAC Name: dipotassium pentachlororutheniumdiuide hydrate
Synonyms: dipotassium pentachlororutheniumdiuide hydrate
Molecular Weight (g/mol): 374.53
Molecular Formula: Cl5H2K2ORu
InChi Key: SRVGBBZEPHUDFZ-UHFFFAOYSA-I
SMILES: O.[K+].[K+].Cl[Ru--](Cl)(Cl)(Cl)Cl
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)