accessibility menu, dialog, popup

UserName

CAS RN 1435-50-3

ClClBr

CAS RN 1435-50-3

IUPAC Name: 2-bromo-1,4-dichlorobenzene
Synonyms: benzene, 2-bromo-1,4-dichloro-
Molecular Weight (g/mol): 225.89
Molecular Formula: C6H3BrCl2
InChi Key: OVXVQBCRONSPDC-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(Br)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results