CAS RN 143158-55-8
CAS RN 143158-55-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-Cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-Cyclopropyl-8-ethoxy-6,7-difluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.