accessibility menu, dialog, popup

UserName

CAS RN 1431329-64-4

CH3OClFFFOCH3

CAS RN 1431329-64-4

IUPAC Name: 4-methoxy-3-methyl-5-(trifluoromethyl)benzoyl chloride
Synonyms: 4-methoxy-3-methyl-5-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 252.62
Molecular Formula: C10H8ClF3O2
InChi Key: YUARQUDWZUWICU-UHFFFAOYSA-N
SMILES: COC1=C(C)C=C(C=C1C(F)(F)F)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results