accessibility menu, dialog, popup

UserName

CAS RN 142878-12-4

H3CO(S)(S)CH3(S)(S)(R)HNNOO

CAS RN 142878-12-4

IUPAC Name: 1-(6-{[(1S,3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]amino}hexyl)pyrrolidine-2,5-dione
Synonyms: 1-(6-{[(1S,3aS,3bR,9bS,11aS)-7-methoxy-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H-cyclopenta[a]phenanthren-1-yl]amino}hexyl)pyrrolidine-2,5-dione
Molecular Weight (g/mol): 466.67
Molecular Formula: C29H42N2O3
InChi Key: CJHWFIUASFBCKN-ZRJUGLEFSA-N
SMILES: COC1=CC=C2[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CCC2=C1)NCCCCCCN1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results