accessibility menu, dialog, popup

UserName

CAS RN 142715-48-8

CH3ONHNNNOCH3OCH3OCH3OCH3OCH3HCl

CAS RN 142715-48-8

IUPAC Name: 4-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-6,7-dimethoxyquinazolin-2-amine hydrochloride
Synonyms: 4-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-6,7-dimethoxyquinazolin-2-amine hydrochloride
Molecular Weight (g/mol): 597.11
Molecular Formula: C31H37ClN4O6
InChi Key: WWCHXVYTCMPAMV-UHFFFAOYSA-N
SMILES: Cl.COC1=CC2=NC(NCCC3=CC(OC)=C(OC)C=C3)=NC(N3CCC4=CC(OC)=C(OC)C=C4C3)=C2C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results