accessibility menu, dialog, popup

UserName

CAS RN 14242-05-8

OClOOOAg

CAS RN 14242-05-8

IUPAC Name: silver(1+) hydrate perchlorate
Synonyms: silver(1+) hydrate perchlorate
Molecular Weight (g/mol): 225.33
Molecular Formula: AgClH2O5
InChi Key: IXFNJWPLHOWEPT-UHFFFAOYSA-M
SMILES: O.[Ag+].[O-][Cl](=O)(=O)=O
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)