accessibility menu, dialog, popup

UserName

HHClClNHHN

CAS RN 142-64-3

IUPAC Name: dihydrogen piperazine dichloride
Synonyms: dihydrogen piperazine dichloride
Molecular Weight (g/mol): 159.05
Molecular Formula: C4H12Cl2N2
InChi Key: CVVIJWRCGSYCMB-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].C1CNCCN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results