accessibility menu, dialog, popup

UserName

CAS RN 141433-60-5

CH3ONOOOH3CCl

CAS RN 141433-60-5

IUPAC Name: 16,17-dimethoxy-5,7-dioxa-13λ⁵-azapentacyclo[11.8.0.0²,¹⁰.0⁴,⁸.0¹⁵,²⁰]henicosa-1(21),2,4(8),9,13,15,17,19-octaen-13-ylium hydrate chloride
Synonyms: berberine hydrate chloride
Molecular Weight (g/mol): 389.83
Molecular Formula: C20H20ClNO5
InChi Key: BPNJXFPOPCFZOC-UHFFFAOYSA-M
SMILES: O.[Cl-].COC1=CC=C2C=C3C4=CC5=C(OCO5)C=C4CC[N+]3=CC2=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)