accessibility menu, dialog, popup

UserName

CAS RN 14017-46-0

LaOClOOOOClOOOOClOOOH2OH2OH2OH2OH2OH2O

CAS RN 14017-46-0

IUPAC Name: lanthanum(3+) triperchlorate
Synonyms: lanthanum(3+) triperchlorate
Molecular Weight (g/mol): 437.24
Molecular Formula: Cl3LaO12
InChi Key: MYGVQOYLIMJKKU-UHFFFAOYSA-K
SMILES: [La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results