CAS RN 1391051-68-5
CAS RN 1391051-68-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-Nitroso Nebivolol (Mixture of Diastereomers), TRC
CAS: 1391051-68-5 Molecular Formula: C22 H24 F2 N2 O5 Molecular Weight (g/mol): 434.43 Synonym: N,N-Bis[2-(6-fluorochroman-2-yl)-2-hydroxyethyl]nitrous amide,N,N-Bis[2-(6-fluorochroman-2-yl)-2-hydroxyethyl]nitrous amide IUPAC Name: N,N-bis[2-(6-fluoro-3,4-dihydro-2H-chromen-2-yl)-2-hydroxyethyl]nitrous amide SMILES: OC(CN(CC(O)C1CCc2cc(F)ccc2O1)N=O)C3CCc4cc(F)ccc4O3
2
N-Nitrosonebivolol, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.