CAS RN 138564-16-6
CAS RN 138564-16-6
IUPAC Name:
2,4-difluoro-N-methylaniline
Synonyms:
2,4-difluoro-N-methylaniline
Molecular Weight (g/mol):
143.14
Molecular Formula:
C7H7F2N
InChi Key:
RPGDVPAENGWKAO-UHFFFAOYSA-N
SMILES:
CNC1=CC=C(F)C=C1F
1
–
1
of
1
results
1
2,4-Difluoro-N-methylaniline, 98%, Thermo Scientific™
CAS: 138564-16-6 Molecular Formula: C7H7F2N Molecular Weight (g/mol): 143.14 MDL Number: MFCD00042472 InChI Key: RPGDVPAENGWKAO-UHFFFAOYSA-N Synonym: n-methyl-2,4-difluoroaniline,n1-methyl-2,4-difluoroaniline,benzenamine,2,4-difluoro-n-methyl,benzenamine, 2,4-difluoro-n-methyl,difluoronmethylaniline,pubchem10041,acmc-20aod4,2,4-difluoro-n-methylaniline,2,4-difluoro-phenyl-methyl-amine,2,4-bis fluoranyl-n-methyl-aniline PubChem CID: 2737023 IUPAC Name: 2,4-difluoro-N-methylaniline SMILES: CNC1=CC=C(F)C=C1F