accessibility menu, dialog, popup

UserName

CAS RN 138500-86-4

CH3H3COBOH3CH3CN

CAS RN 138500-86-4

IUPAC Name: 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
Synonyms: 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
Molecular Weight (g/mol): 243.11
Molecular Formula: C14H18BNO2
InChi Key: URWMFRYGXSHPRV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CC#N)C=C1
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results