CAS RN 13813-24-6
CAS RN 13813-24-6
IUPAC Name:
neodymium(3+) triiodide
Synonyms:
neodymium(3+) triiodide
Molecular Weight (g/mol):
524.96
Molecular Formula:
I3Nd
InChi Key:
DKSXWSAKLYQPQE-UHFFFAOYSA-K
SMILES:
[I-].[I-].[I-].[Nd+3]
1
–
1
of
1
results
1
Neodymium(III) iodide, ultra dry, 99.9% (metals basis), Thermo Scientific™
CAS: 13813-24-6 MDL Number: MFCD00016248