accessibility menu, dialog, popup

UserName

CAS RN 1373920-85-4

CH3OOOHFFFF

CAS RN 1373920-85-4

IUPAC Name: 2-[2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-fluoro-5-methoxy-3-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 252.17
Molecular Formula: C10H8F4O3
InChi Key: LDYVUXOKNKIQKV-UHFFFAOYSA-N
SMILES: COC1=CC(=C(F)C(CC(O)=O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results