accessibility menu, dialog, popup

UserName

CAS RN 13710-19-5

CH3HNOHOCl

CAS RN 13710-19-5

IUPAC Name: 2-[(4-chloro-3-methylphenyl)amino]benzoic acid
Synonyms: clotam acido tolfenamico pharmacia brand of tolfenamic acid bifenac provalis brand of tolfenamic acid zambon brand of tolfenamic acid tolfenamic acid (product) acide tolfenamique tolfenamic acid (substance) rociclyn
Molecular Weight (g/mol): 261.71
Molecular Formula: C14H12ClNO2
InChi Key: QDNMBJXNLJFNHT-UHFFFAOYSA-N
SMILES: CC1=CC(NC2=CC=CC=C2C(O)=O)=CC=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Concentration 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)