accessibility menu, dialog, popup

UserName

CAS RN 13569-63-6

ClClClRe

CAS RN 13569-63-6

IUPAC Name: rhenium(3+) trichloride
Synonyms: trirhenium nonachloride
Molecular Weight (g/mol): 292.56
Molecular Formula: Cl3Re
InChi Key: LOIHSHVELSAXQN-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Re+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results