CAS RN 13551-90-1
CAS RN 13551-90-1
IUPAC Name:
2-nitro-1-[(oxiran-2-yl)methyl]-1H-imidazole
Synonyms:
2-nitro-1-(oxiran-2-ylmethyl)imidazole
Molecular Weight (g/mol):
169.14
Molecular Formula:
C6H7N3O3
InChi Key:
SYFMSLVOHQZYEB-UHFFFAOYNA-N
SMILES:
[O-][N+](=O)C1=NC=CN1CC1CO1
1
–
2
of
2
results
1
1-(2,3-Epoxypropyl)-2-nitroimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(2,3-Epoxypropyl)-2-nitroimidazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.