accessibility menu, dialog, popup

UserName

H2OH2OH2OH2OH2OH2OYONOOONOOONOO

CAS RN 13494-98-9

IUPAC Name: yttrium(3+) hexahydrate trinitrate
Synonyms: yttrium(3+) hexahydrate trinitrate
Molecular Weight (g/mol): 383.01
Molecular Formula: H12N3O15Y
InChi Key: QBAZWXKSCUESGU-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)