CAS RN 1346602-13-8
CAS RN 1346602-13-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carteolol-d9 Hydrochloride, TRC
CAS: 1346602-13-8 Molecular Formula: C16H16D9ClN2O3 Molecular Weight (g/mol): 337.89 Synonym: 5-[3-[(1,1-Dimethylethyl-d9)amino]-2-hydroxypropoxy]-3,4-dihydro-2(1H)-quinolinone Hydrochloride,(+/-)-Carteolol-d9 Hydrochloride,dl-Carteolol-d9 Hydrochloride,Abbott 43326-d9,OPC-1085-d9,Arteoptic-d9,Endak-d9,Mikelan-d9,Teoptic-d9 IUPAC Name: 5-[3-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]-2-hydroxypropoxy]-3,4-dihydro-1H-quinolin-2-one;hydrochloride SMILES: O=C1CCC2=C(OCC(O)CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C=CC=C2N1.Cl