CAS RN 1332605-57-8
CAS RN 1332605-57-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(S)-4-Hydroxy-4-methyl-7,8-dihydro-1H-pyrano[3,4-f]-indolizine-3,6,10(4H)-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(S)-4-Hydroxy-4-methyl-7,8-dihydro-1H-pyrano[3,4-f]-indolizine-3,6,10(4H)-trione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.